missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindazole-6-boronic acid, 97%

Catalog No. AC430480010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430480010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430480010 Supplier Thermo Scientific Chemicals Supplier No. 430480010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1150114-80-9 | C8H9BN2O2 | 175.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1150114-80-9
Molecular Formula C8H9BN2O2
Molecular Weight (g/mol) 175.98
MDL Number MFCD09870053
InChI Key OMVQAWCHQBKZCF-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-6-yl boronic acid, 1-methyl-1h-indazole-6-boronic acid, 1-methylindazole-6-boronic acid, 1-methyl-1h-indazol-6-boronic acid, 1-methyl-1h-indazol-6-ylboronic acid, 6-borono-1-methyl-1h-indazole, 1-methyl-1h-indazol-6-yl-6-boronic acid, pubchem20896, acmc-2099ne, 1-methyl-indazole-6-boronic acid
PubChem CID 44118215
IUPAC Name (1-methylindazol-6-yl)boronic acid
SMILES CN1N=CC2=CC=C(C=C12)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 259.8°C to 261.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 175.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylindazole-6-boronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only