missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylindazole-3-carboxylic Acid 98.0+%, TCI America™
$156.02 - $466.02
Chemical Identifiers
| CAS | 50890-83-0 |
|---|---|
| Molecular Formula | C9H8N2O2 |
| Molecular Weight (g/mol) | 176.175 |
| MDL Number | MFCD00272569 |
| InChI Key | OVVDFORZEGKEJM-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid |
| PubChem CID | 689105 |
| IUPAC Name | 1-methylindazole-3-carboxylic acid |
| SMILES | CN1C2=CC=CC=C2C(=N1)C(=O)O |
Chemical Identifiers
| 50890-83-0 | |
| 176.175 | |
| OVVDFORZEGKEJM-UHFFFAOYSA-N | |
| 689105 | |
| CN1C2=CC=CC=C2C(=N1)C(=O)O |
| C9H8N2O2 | |
| MFCD00272569 | |
| 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid | |
| 1-methylindazole-3-carboxylic acid |
Specifications
| 50890-83-0 | |
| White | |
| MFCD00272569 | |
| 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid | |
| CN1C2=CC=CC=C2C(=N1)C(=O)O | |
| 176.175 | |
| 176.18 | |
| Crystalline Powder |
| 216°C | |
| C9H8N2O2 | |
| 1 g | |
| OVVDFORZEGKEJM-UHFFFAOYSA-N | |
| 1-methylindazole-3-carboxylic acid | |
| 689105 | |
| ≥98.0% (GC,T) | |
| 1-Methylindazole-3-carboxylic Acid |
Safety and Handling
TSCA : No