missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindazole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. M21435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21435G 5 g
M21431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21435G Supplier TCI America Supplier No. M21435G

Chemical Identifiers

CAS 50890-83-0
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00272569
InChI Key OVVDFORZEGKEJM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid
PubChem CID 689105
IUPAC Name 1-methylindazole-3-carboxylic acid
SMILES CN1C2=CC=CC=C2C(=N1)C(=O)O

Specifications

Melting Point 216°C
Color White
Quantity 5 g
Formula Weight 176.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylindazole-3-carboxylic Acid