missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylindazole-3-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 216°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 176.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Methylindazole-3-carboxylic Acid |
Safety and Handling
| TSCA | No |