Learn More
1-Methylindazole-3-carboxylic acid, 97%
CAS: 50890-83-0 | C9H8N2O2 | 176.175 g/mol
$67.96 - $206.53
Chemical Identifiers
| CAS | 50890-83-0 |
|---|---|
| Molecular Formula | C9H8N2O2 |
| Molecular Weight (g/mol) | 176.175 |
| MDL Number | MFCD00272569 |
| InChI Key | OVVDFORZEGKEJM-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid |
| PubChem CID | 689105 |
| IUPAC Name | 1-methylindazole-3-carboxylic acid |
| SMILES | CN1C2=CC=CC=C2C(=N1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2687903
|
Thermo Scientific Chemicals
H2687903 |
1 g |
N/A
|
|
|||||
|
AAH2687906
|
Thermo Scientific Chemicals
H2687906 |
5 g |
N/A
|
|
|||||
Description
1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.
Solubility
Insoluble in water. Soluble in DMSO and Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Chemical Identifiers
| 50890-83-0 | |
| 176.175 | |
| OVVDFORZEGKEJM-UHFFFAOYSA-N | |
| 689105 | |
| CN1C2=CC=CC=C2C(=N1)C(=O)O |
| C9H8N2O2 | |
| MFCD00272569 | |
| 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid | |
| 1-methylindazole-3-carboxylic acid |
Specifications
| 50890-83-0 | |
| C9H8N2O2 | |
| 1 g | |
| Insoluble in water. Soluble in DMSO and Methanol. | |
| CN1C2=CC=CC=C2C(=N1)C(=O)O | |
| 176.175 | |
| 176.17 | |
| 1-Methylindazole-3-carboxylic acid |
| ∽218°C (decomposition) | |
| MFCD00272569 | |
| 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid | |
| OVVDFORZEGKEJM-UHFFFAOYSA-N | |
| 1-methylindazole-3-carboxylic acid | |
| 689105 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only