missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindazole-3-carboxylic acid, 97%

Catalog No. AAH2687906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2687906 5 g
AAH2687903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2687906 Supplier Thermo Scientific Chemicals Supplier No. H2687906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50890-83-0 | C9H8N2O2 | 176.175 g/mol

1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

Solubility
Insoluble in water. Soluble in DMSO and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 50890-83-0
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00272569
InChI Key OVVDFORZEGKEJM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid
PubChem CID 689105
IUPAC Name 1-methylindazole-3-carboxylic acid
SMILES CN1C2=CC=CC=C2C(=N1)C(=O)O

Specifications

Melting Point ∽218°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water. Soluble in DMSO and Methanol.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 1-Methylindazole-3-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only