missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole 99.0+%, TCI America™

Catalog No. M0508500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0508500G 500 g
M050825G 25 g
M0508100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0508500G Supplier TCI America Supplier No. M0508500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
PubChem CID 1390
ChEBI CHEBI:113454
IUPAC Name 1-methyl-1H-imidazole
SMILES CN1C=CN=C1

Specifications

Color Yellow
pH 11.3 (100g/L H2O soln.)
Boiling Point 198°C
Quantity 500 g
UN Number 1760
Formula Weight 82.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylimidazole
EINECSNumber (5)-5813
RTECSNumber NI7000000
TSCA Yes