missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole 99.0+%, TCI America™

Supplier:  TCI America M0508100G

 View more versions of this product

Catalog No. M0508100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Methylimidazole
Yellow
198°C
MFCD00005292
1760
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1-methyl-1H-imidazole
1390
82.11
Liquid
616-47-7
11.3 (100g/L H2O soln.)
C4H6N2
100 g
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
CN1C=CN=C1
82.11
CHEBI:113454
≥99.0% (GC)

Chemical Identifiers

616-47-7
82.11
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1390
1-methyl-1H-imidazole
C4H6N2
MFCD00005292
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
CHEBI:113454
CN1C=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5813

RTECSNumber : NI7000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions