missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 1-Methylimidazole, for biochemistry, anhydrous, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -2.0°C |
| Density | 1.0300 g/mL |
| Boiling Point | 198.0°C |
| Flash Point | 92°C |
| Refractive Index | 1.4955 to 1.4975 |
| Quantity | 1 L |
| Specific Gravity | 1.03 |
| Solubility Information | Solubility aq. soln.: 145 g/L (25°C). Other solubilities: soluble in organic solvents |
| Formula Weight | 82.12 |
| Grade | Biochemistry |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately call a POISON CENTER or doctor/physician. |
| EINECSNumber | 210-484-7 |
RUO – Research Use Only