missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1-Methylimidazole, for biochemistry, anhydrous, AcroSeal™

Catalog No. AC465290025
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465290025 2.5 L
AC465290010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465290025 Supplier Thermo Scientific Chemicals Supplier No. 465290025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
IUPAC Name 1-methyl-1H-imidazole
SMILES CN1C=CN=C1

Specifications

Melting Point -2.0°C
Density 1.0300 g/mL
Boiling Point 198.0°C
Flash Point 92°C
Refractive Index 1.4955 to 1.4975
Quantity 2.5 L
Specific Gravity 1.03
Solubility Information Solubility aq. soln.: 145 g/L (25°C). Other solubilities: soluble in organic solvents
Formula Weight 82.12
Grade Biochemistry
Chemical Name or Material 1-Methylimidazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Toxic in contact with skin.

Causes severe skin burns and eye damage.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF ON SKIN: Wash with plenty of soap and water.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Do not breathe dust/fume/gas/mist/vapors/spray.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 210-484-7

RUO – Research Use Only