missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-Methylimidazole

Catalog No. M50834500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M50834500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M50834500G Supplier Sigma Aldrich Supplier No. M50834500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methylimidazole

Specifications

CAS 616-47-7
Melting Point -6°C (lit.)
Density 1.03 g/mL (at 25°C (literature))
Boiling Point 198°C (lit.)
Molecular Formula C4H6N2
Refractive Index n20/D 1.495 (literature)
Linear Formula C4H6N2
MDL Number MFCD00005292
Quantity 500 g
Molecular Weight (g/mol) 82.1
Percent Purity 99%
Show More Show Less