missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-Methylimidazole

Catalog No. 3360922L
Encompass
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3360922L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3360922L Supplier Sigma Aldrich Supplier No. 3360922L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methylimidazole

Specifications

CAS 616-47-7
Melting Point -6°C
Density 1.03 g/mL at 25°C, 1.034-1.036 g/mL
Boiling Point 198°C
Molecular Formula C4H6N2
Refractive Index n20/D 1.495
Linear Formula C4H6N2
MDL Number MFCD00005292
Quantity 2 L
Molecular Weight (g/mol) 82.1 g/mol