missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. M21761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21761G 1 g
M21765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21761G Supplier TCI America Supplier No. M21761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13750-81-7
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01321308
InChI Key UEBFLTZXUXZPJO-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6
PubChem CID 139575
IUPAC Name 1-methyl-1H-imidazole-2-carbaldehyde
SMILES CN1C=CN=C1C=O

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 74°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Methylimidazole-2-carboxaldehyde