missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylcyclopentanol 98.0+%, TCI America™
$158.44 - $303.84
Chemical Identifiers
| CAS | 1462-03-9 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00001364 |
| InChI Key | CAKWRXVKWGUISE-UHFFFAOYSA-N |
| PubChem CID | 73830 |
| IUPAC Name | 1-methylcyclopentan-1-ol |
| SMILES | CC1(CCCC1)O |
Chemical Identifiers
| 1462-03-9 | |
| 100.161 | |
| CAKWRXVKWGUISE-UHFFFAOYSA-N | |
| 1-methylcyclopentan-1-ol |
| C6H12O | |
| MFCD00001364 | |
| 73830 | |
| CC1(CCCC1)O |
Specifications
| 1462-03-9 | |
| C6H12O | |
| 10 g | |
| 1-methylcyclopentanol, 1-methyl cyclopentanol, cyclopentanol, 1-methyl, 1-methyl-cyclopentanol, 1-hydroxy-1-methylcyclopentane, cyclopentanol, methyl, 1-methyl-1-cyclopentanol, 1-methylcyclopentanolc, cyclopentanol,1-methyl | |
| CC1(CCCC1)O | |
| 100.161 | |
| 100.16 | |
| Crystals |
| White | |
| MFCD00001364 | |
| 1325 | |
| CAKWRXVKWGUISE-UHFFFAOYSA-N | |
| 1-methylcyclopentan-1-ol | |
| 73830 | |
| ≥98.0% (GC) | |
| 1-Methylcyclopentanol |
Safety and Handling
TSCA : No