missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentanol 98.0+%, TCI America™

Catalog No. M072910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M072910G 10 g
M072925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M072910G Supplier TCI America Supplier No. M072910G
Only null left

Chemical Identifiers

CAS 1462-03-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001364
InChI Key CAKWRXVKWGUISE-UHFFFAOYSA-N
PubChem CID 73830
IUPAC Name 1-methylcyclopentan-1-ol
SMILES CC1(CCCC1)O

Specifications

Color White
Quantity 10 g
UN Number 1325
Synonym 1-methylcyclopentanol, 1-methyl cyclopentanol, cyclopentanol, 1-methyl, 1-methyl-cyclopentanol, 1-hydroxy-1-methylcyclopentane, cyclopentanol, methyl, 1-methyl-1-cyclopentanol, 1-methylcyclopentanolc, cyclopentanol,1-methyl
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1-Methylcyclopentanol