missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentanol, 98%

Catalog No. AAB2474406
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2474406 5 g
AAB2474422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2474406 Supplier Thermo Scientific Chemicals Supplier No. B2474406
Only null left

CAS: 1462-03-9 | C6H12O | 100.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1462-03-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001364
InChI Key CAKWRXVKWGUISE-UHFFFAOYSA-N
PubChem CID 73830
IUPAC Name 1-methylcyclopentan-1-ol
SMILES CC1(CCCC1)O

Specifications

Melting Point 36°C to 37°C
Density 0.904
Boiling Point 135°C to 136°C
Flash Point 40°C (105°F)
Quantity 5 g
UN Number UN1325
Synonym 1-methylcyclopentanol, 1-methyl cyclopentanol, cyclopentanol, 1-methyl, 1-methyl-cyclopentanol, 1-hydroxy-1-methylcyclopentane, cyclopentanol, methyl, 1-methyl-1-cyclopentanol, 1-methylcyclopentanolc, cyclopentanol,1-methyl
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopentanol
Hazard Category H228
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 215-963-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only