missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanol 97.0+%, TCI America™

Catalog No. M032825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M032825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M032825ML Supplier TCI America Supplier No. M032825ML

Chemical Identifiers

CAS 590-67-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00003857
InChI Key VTBOTOBFGSVRMA-UHFFFAOYSA-N
Synonym 1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol
PubChem CID 11550
IUPAC Name 1-methylcyclohexan-1-ol
SMILES CC1(CCCCC1)O

Specifications

Color White
Boiling Point 155°C
Quantity 25 mL
Formula Weight 114.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Methylcyclohexanol
EINECSNumber (3)-2301
TSCA No