missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanol, 96%

CAS: 590-67-0 | C7H14O | 114.19 g/mol

$204.32 - $204.32

Chemical Identifiers

CAS 590-67-0
MDL Number MFCD00003857
InChI Key VTBOTOBFGSVRMA-UHFFFAOYSA-N
Synonym 1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol
PubChem CID 11550
IUPAC Name 1-methylcyclohexan-1-ol
SMILES CC1(CCCCC1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126600250
SDS View Documents
Thermo Scientific Chemicals
126600250
25 g Glass bottle
Each for $204.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

590-67-0
VTBOTOBFGSVRMA-UHFFFAOYSA-N
11550
CC1(CCCCC1)O
MFCD00003857
1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol
1-methylcyclohexan-1-ol

Specifications

590-67-0
100.0
0.9190g/mL
96%
C7H14O
CH3C6H10OH
25 g
1-methylcyclohexanol, cyclohexanol, 1-methyl, 1-methyl-1-cyclohexanol, cyclohexanol,methyl, 1-methyl-cyclohexanol, 1-methylcyclohexyl alcohol, acmc-1b1ai, 1-methyl-cyclohexan-1-ol, 1-methyl cyclohexanol
CC1(CCCCC1)O
114.188
114.19
Liquid After Melting
95.0
Colorless to Yellow
Authentic
Glass bottle
1.4590 to 1.4610 (20°C, 589nm)
MFCD00003857
06, 11
VTBOTOBFGSVRMA-UHFFFAOYSA-N
1-methylcyclohexan-1-ol
11550
≥95%
1-Methylcyclohexanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only