missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylbenzimidazole 98.0+%, TCI America™
$126.76 - $434.32
Chemical Identifiers
| CAS | 1632-83-3 |
|---|---|
| Molecular Formula | C8H8N2 |
| Molecular Weight (g/mol) | 132.17 |
| MDL Number | MFCD00192275 |
| InChI Key | FGYADSCZTQOAFK-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole |
| PubChem CID | 95890 |
| IUPAC Name | 1-methyl-1H-1,3-benzodiazole |
| SMILES | CN1C=NC2=CC=CC=C12 |
Chemical Identifiers
| 1632-83-3 | |
| 132.17 | |
| FGYADSCZTQOAFK-UHFFFAOYSA-N | |
| 95890 | |
| CN1C=NC2=CC=CC=C12 |
| C8H8N2 | |
| MFCD00192275 | |
| 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole | |
| 1-methyl-1H-1,3-benzodiazole |
Specifications
| 1632-83-3 | |
| 154°C | |
| MFCD00192275 | |
| 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole | |
| CN1C=NC2=CC=CC=C12 | |
| 132.17 | |
| 132.17 | |
| Crystalline Powder |
| 59°C | |
| C8H8N2 | |
| 5 g | |
| FGYADSCZTQOAFK-UHFFFAOYSA-N | |
| 1-methyl-1H-1,3-benzodiazole | |
| 95890 | |
| ≥98.0% (GC,T) | |
| 1-Methylbenzimidazole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator