missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbenzimidazole 98.0+%, TCI America™

Catalog No. M21735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21735G 5 g
M217325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21735G Supplier TCI America Supplier No. M21735G

Chemical Identifiers

CAS 1632-83-3
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00192275
InChI Key FGYADSCZTQOAFK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole
PubChem CID 95890
IUPAC Name 1-methyl-1H-1,3-benzodiazole
SMILES CN1C=NC2=CC=CC=C12

Specifications

Melting Point 59°C
Boiling Point 154°C
Quantity 5 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylbenzimidazole
TSCA No
Recommended Storage Refrigerator