missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbenzimidazole, 97%

Catalog No. AAB2547206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2547206 5 g
AAB2547214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2547206 Supplier Thermo Scientific Chemicals Supplier No. B2547206

CAS: 1632-83-3 | C8H8N2 | 132.17 g/mol

1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 1632-83-3
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00192275
InChI Key FGYADSCZTQOAFK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole
PubChem CID 95890
IUPAC Name 1-methylbenzimidazole
SMILES CN1C=NC2=CC=CC=C12

Specifications

Melting Point 59°C to 62°C
Boiling Point 154°C (12 mmHg)
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 132.17
Percent Purity 99%
Chemical Name or Material 1-Methylbenzimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only