missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1-Methyladenine, 95%, Thermo Scientific Chemicals

Catalog No. AC201310250
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201310250 25 mg Glass bottle
AC201311000 100 mg Glass bottle
AC201312500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC201310250 Supplier Thermo Scientific Chemicals Supplier No. 201310250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5142-22-3 | C6H7N5 | 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5142-22-3
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00010532
InChI Key HPZMWTNATZPBIH-UHFFFAOYSA-N
Synonym 1-methyladenine, 1-methyl-1h-purin-6-amine, 1h-purin-6-amine, 1-methyl, adenine, 1-methyl, n1-methyladenine, 1-methyl-1h-purin-6-ylamine, 6h-purin-6-imine, 1,9-dihydro-1-methyl, 1-methyl-adenine, 1-methyl-9h-purin-6-imine, 1-methylhydropurine-6-ylamine
PubChem CID 78821
ChEBI CHEBI:18083
IUPAC Name 1-methylpurin-6-amine
SMILES CN1C=NC2=NC=NC2=C1N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point >300.0°C
Color Beige or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 mg
Beilstein 26, V,17, 136
Solubility Information Solubility in water: soluble
Formula Weight 149.15
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyladenine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-907-

RUO – Research Use Only