missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™

Catalog No. p-7132938
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M19611G 1 g
M19615G 5 g
2 options

Catalog No. M19611G

Supplier: TCI America M19611G

Only null left

Chemical Identifiers

CAS 5672-83-3
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.291
MDL Number MFCD00083278
InChI Key BGMCTGARFXPQML-NSHDSACASA-N
Synonym z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester
PubChem CID 2733408
IUPAC Name (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 71°C
Color White
Quantity 1 g
Formula Weight 295.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl N-Carbobenzoxy-L-glutamate