missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl N-Carbobenzoxy-L-aspartate 98.0+%, TCI America™
$184.54 - $726.98
Chemical Identifiers
| CAS | 4668-42-2 |
|---|---|
| Molecular Formula | C13H15NO6 |
| Molecular Weight (g/mol) | 281.264 |
| MDL Number | MFCD00083277 |
| InChI Key | MFFFBNAPQRDRQW-JTQLQIEISA-N |
| Synonym | z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988 |
| PubChem CID | 7021783 |
| IUPAC Name | (3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid |
| SMILES | COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 4668-42-2 | |
| 281.264 | |
| MFFFBNAPQRDRQW-JTQLQIEISA-N | |
| 7021783 | |
| COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1 |
| C13H15NO6 | |
| MFCD00083277 | |
| z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988 | |
| (3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
| 4668-42-2 | |
| White | |
| MFCD00083277 | |
| z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988 | |
| COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 281.264 | |
| 281.26 | |
| Crystalline Powder |
| 91°C | |
| C13H15NO6 | |
| 5 g | |
| MFFFBNAPQRDRQW-JTQLQIEISA-N | |
| (3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid | |
| 7021783 | |
| ≥98.0% (HPLC,T) | |
| 1-Methyl N-Carbobenzoxy-L-aspartate |
Safety and Handling
TSCA : No