missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific™

Catalog No. AAH5140503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5140503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5140503 Supplier Thermo Scientific Chemicals Supplier No. H5140503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Methyl-5-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 147716-03-8
Molecular Formula C16H32N2Sn
Molecular Weight (g/mol) 371.156
MDL Number MFCD01319031
InChI Key OGYWKJKAIAEDQX-UHFFFAOYSA-N
PubChem CID 10915651
IUPAC Name tributyl-(3-methylimidazol-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CN1C

Specifications

Quantity 1 g
UN Number UN2788
Synonym 1-methyl-5-tributylstannyl-1h-imidazole, 1-methyl-5-tributylstannanyl-1h-imidazole, 1-methyl-5-tributylstannyl imidazole, tributyl-3-methylimidazol-4-yl stannane, 1h-imidazole,1-methyl-5-tributylstannyl, 1h-imidazole, 1-methyl-5-tributylstannyl, pubchem22530, acmc-1cabs, 1-methyl-5-tributylstannylimidazole, 5-tributylstannyl-1-methylimidazole
Solubility Information Sparingly soluble in water.
Formula Weight 371.15
Percent Purity ≥90%
Chemical Name or Material 1-Methyl-5-(tri-n-butylstannyl)imidazole
Hazard Category H301-H312-H315-H319-H360FD-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures