missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC47101DA

Quantity 1g

 View more versions of this product

Catalog No. CC47101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

869901-15-5
208.235
URCWVEMABYSGJA-UHFFFAOYSA-N
7537648
CN1C(=CC(=N1)C(=O)O)C2=CC=CS2
C9H8N2O2S
MFCD08271952
1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl, 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl, 1-methyl-5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-5-thien-2-yl-1h-pyrazole
1-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid

Specifications

1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid
96.5
97%
C9H8N2O2S
MFCD08271952
URCWVEMABYSGJA-UHFFFAOYSA-N
1-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
7537648
97%
869901-15-5
100.0
Amber Glass Bottle
1g
1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl, 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl, 1-methyl-5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-5-thien-2-yl-1h-pyrazole
CN1C(=CC(=N1)C(=O)O)C2=CC=CS2
208.235
208.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents