missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC47101CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC47101CB 250mg
CC47101DA 1g
CC47101DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC47101CB Supplier Thermo Scientific Chemicals Supplier No. CC47101CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 869901-15-5
Molecular Formula C9H8N2O2S
Molecular Weight (g/mol) 208.235
MDL Number MFCD08271952
InChI Key URCWVEMABYSGJA-UHFFFAOYSA-N
Synonym 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl, 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl, 1-methyl-5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-5-thien-2-yl-1h-pyrazole
PubChem CID 7537648
IUPAC Name 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
SMILES CN1C(=CC(=N1)C(=O)O)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.