missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

$392.48 - $392.48

Chemical Identifiers

CAS 105994-75-0
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD04038957
InChI Key MXGOKNNGBUOAGF-UHFFFAOYSA-N
Synonym 1-methyl-5-phenyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-methyl-5-phenyl, acmc-1c8ot, 1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenyl, 1ff
PubChem CID 2795474
IUPAC Name 1-methyl-5-phenylpyrazole-4-carboxylic acid
SMILES CN1C(=C(C=N1)C(=O)O)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC28101DA
SDS View Documents
Thermo Scientific Chemicals
CC28101DA
1 g Amber Glass Bottle
Each for $392.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

105994-75-0
202.213
MXGOKNNGBUOAGF-UHFFFAOYSA-N
2795474
CN1C(=C(C=N1)C(=O)O)C2=CC=CC=C2
C11H10N2O2
MFCD04038957
1-methyl-5-phenyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-methyl-5-phenyl, acmc-1c8ot, 1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenyl, 1ff
1-methyl-5-phenylpyrazole-4-carboxylic acid

Specifications

105994-75-0
100.0
Amber Glass Bottle
MFCD04038957
1-methyl-5-phenyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-methyl-5-phenyl, acmc-1c8ot, 1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenyl, 1ff
CN1C(=C(C=N1)C(=O)O)C2=CC=CC=C2
202.213
202.21
1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid
96.5
97%
C11H10N2O2
1 g
MXGOKNNGBUOAGF-UHFFFAOYSA-N
1-methyl-5-phenylpyrazole-4-carboxylic acid
2795474
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'