missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. M26815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26815G 5 g
M26811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26815G Supplier TCI America Supplier No. M26815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 847818-74-0
Molecular Formula C10H17BN2O2
Molecular Weight (g/mol) 208.07
MDL Number MFCD05861380
InChI Key HLXOVAMYQUFLPE-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-5-boronic acid pinacol ester, cbi-bb zero/005567, 1h-pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 4,4,5,5-tetramethyl-2-1-methylpyrazol-5-yl-1,3,2-dioxaborolane
PubChem CID 4912908
IUPAC Name 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CN1N=CC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 71°C
Color White
Quantity 5 g
Formula Weight 208.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole