missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine Hydrochloride 98.0+%, TCI America™

Catalog No. M269025MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M269025MG 25 mg
M2690100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M269025MG Supplier TCI America Supplier No. M269025MG

Chemical Identifiers

CAS 23007-85-4
Molecular Formula C12H16ClN
Molecular Weight (g/mol) 209.72
MDL Number MFCD00012790
InChI Key KOWJANGMTAZWDT-UHFFFAOYSA-N
Synonym MPTP Hydrochloride
PubChem CID 161406
IUPAC Name hydrogen 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine chloride
SMILES [H+].[Cl-].CN1CCC(=CC1)C1=CC=CC=C1

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 209.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine Hydrochloride
RTECSNumber UT8358950
TSCA No