missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide 96.0+%, TCI America™
$242.33 - $835.10
Chemical Identifiers
| CAS | 139756-01-7 |
|---|---|
| Molecular Formula | C8H12N4O3 |
| Molecular Weight (g/mol) | 212.209 |
| MDL Number | MFCD02253196 |
| InChI Key | BMLPAJIEDKJHSB-UHFFFAOYSA-N |
| Synonym | 5-Carbamoyl-1-methyl-4-nitro-3-propylpyrazole |
| PubChem CID | 2754492 |
| IUPAC Name | 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide |
| SMILES | CCCC1=NN(C(=C1[N+](=O)[O-])C(=O)N)C |
Chemical Identifiers
| 139756-01-7 | |
| 212.209 | |
| BMLPAJIEDKJHSB-UHFFFAOYSA-N | |
| 2754492 | |
| CCCC1=NN(C(=C1[N+](=O)[O-])C(=O)N)C |
| C8H12N4O3 | |
| MFCD02253196 | |
| 5-Carbamoyl-1-methyl-4-nitro-3-propylpyrazole | |
| 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide |
Specifications
| 139756-01-7 | |
| C8H12N4O3 | |
| 5 g | |
| BMLPAJIEDKJHSB-UHFFFAOYSA-N | |
| 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide | |
| 2754492 | |
| ≥96.0% (GC) | |
| 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide |
| 139°C | |
| MFCD02253196 | |
| 5-Carbamoyl-1-methyl-4-nitro-3-propylpyrazole | |
| CCCC1=NN(C(=C1[N+](=O)[O-])C(=O)N)C | |
| 212.209 | |
| 212.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No