missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide 96.0+%, TCI America™

Catalog No. M27735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27735G 5 g
M277325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27735G Supplier TCI America Supplier No. M27735G
Only null left

Chemical Identifiers

CAS 139756-01-7
Molecular Formula C8H12N4O3
Molecular Weight (g/mol) 212.209
MDL Number MFCD02253196
InChI Key BMLPAJIEDKJHSB-UHFFFAOYSA-N
Synonym 5-Carbamoyl-1-methyl-4-nitro-3-propylpyrazole
PubChem CID 2754492
IUPAC Name 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide
SMILES CCCC1=NN(C(=C1[N+](=O)[O-])C(=O)N)C

Specifications

Melting Point 139°C
Quantity 5 g
Formula Weight 212.21
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide