missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-imidazolecarboxylic Acid 98.0+%, TCI America™

Catalog No. M22725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22725G 5 g
M22721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22725G Supplier TCI America Supplier No. M22725G

Chemical Identifiers

CAS 41716-18-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD02179560
InChI Key WZTRQGJMMHMFGH-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylicacid, pubchem9001, acmc-1am6x, 1-methyl-4-imidazolic acid, ksc237q3j, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-1h-4carboxylic acid
PubChem CID 541509
ChEBI CHEBI:74746
IUPAC Name 1-methylimidazole-4-carboxylic acid
SMILES CN1C=C(N=C1)C(=O)O

Specifications

Melting Point 247°C
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-imidazolecarboxylic Acid