missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. p-7133168
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M23391G 1 g
M23395G 5 g
2 options

Catalog No. M23391G

Supplier: TCI America M23391G

Only null left

Chemical Identifiers

CAS 761446-44-0
Molecular Formula C10H17BN2O2
Molecular Weight (g/mol) 208.07
MDL Number MFCD03789259
InChI Key UCNGGGYMLHAMJG-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-4-pyrazoleboronic acid pinacol ester
PubChem CID 2773987
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole