missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 97%, Thermo Scientific™

Catalog No. CC45839CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45839CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC45839CB Supplier Thermo Scientific Chemicals Supplier No. CC45839CB

Chemical Identifiers

CAS 898289-06-0
Molecular Formula C15H20BNO2
Molecular Weight (g/mol) 257.14
MDL Number MFCD08690266
InChI Key CBEYYYCGOCYJIK-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1h-indole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methylindol-4-boronic acid pinacol ester, 1-methylindol-4-boronic acid, pinacol ester, 1-methyl-1h-indol-4-yl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindol-4-yl-1,3,2-dioxaborolane, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-indole
PubChem CID 18525785
SMILES CN1C=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Formula Weight 257.14
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.