missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(3-nitrophenyl)piperazine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 148546-97-8
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD08435858
InChI Key IIRKKCDXJIXWHI-UHFFFAOYSA-N
Synonym 1-methyl-4-3-nitrophenyl piperazine, 4-methyl-1-3-nitrophenyl piperazine, piperazine,1-methyl-4-3-nitrophenyl, zlchem 196, pubchem18685, acmc-209d0u, 1-3-nitrophenyl 4-methylpiperazine, 1-3-nitrophenyl-4-methylpiperazine, 1-methyl-4-3-nitrophenyl-piperazine, 1-methyl-4-3-nitro-phenyl-piperazine
PubChem CID 12152161
IUPAC Name 1-methyl-4-(3-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC35925DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC35925DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

148546-97-8
221.26
IIRKKCDXJIXWHI-UHFFFAOYSA-N
12152161
CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O
C11H15N3O2
MFCD08435858
1-methyl-4-3-nitrophenyl piperazine, 4-methyl-1-3-nitrophenyl piperazine, piperazine,1-methyl-4-3-nitrophenyl, zlchem 196, pubchem18685, acmc-209d0u, 1-3-nitrophenyl 4-methylpiperazine, 1-3-nitrophenyl-4-methylpiperazine, 1-methyl-4-3-nitrophenyl-piperazine, 1-methyl-4-3-nitro-phenyl-piperazine
1-methyl-4-(3-nitrophenyl)piperazine

Specifications

148546-97-8
100.0
Amber Glass Bottle
MFCD08435858
IIRKKCDXJIXWHI-UHFFFAOYSA-N
1-methyl-4-(3-nitrophenyl)piperazine
12152161
97%
96.5
97%
C11H15N3O2
1-methyl-4-3-nitrophenyl piperazine, 4-methyl-1-3-nitrophenyl piperazine, piperazine,1-methyl-4-3-nitrophenyl, zlchem 196, pubchem18685, acmc-209d0u, 1-3-nitrophenyl 4-methylpiperazine, 1-3-nitrophenyl-4-methylpiperazine, 1-methyl-4-3-nitrophenyl-piperazine, 1-methyl-4-3-nitro-phenyl-piperazine
CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O
221.26
221.26
1-Methyl-4-(3-nitrophenyl)piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents