missing translation for 'onlineSavingsMsg'
Learn More

1-Méthyl-3-(trifluorométhyl)pyrazole-4-carboxylique Acide 98,0+%, TCI America™

Numéro de catalogue. M26281G
Modifier l'affichage
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M26281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M26281G Fournisseur TCI America Code fournisseur M26281G

Identifiants chimiques

CAS 113100-53-1
Molecular Formula C6H5F3N2O2
Molecular Weight (g/mol) 194.113
MDL Number MFCD01936005
InChI Key FZNKJQNEJGXCJH-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 3-trifluoromethyl-1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl-4-pyrazolecarboxylic acid, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxy, 1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl, pubchem11402, acmc-1bscj, ksc183q3j
PubChem CID 2775665
IUPAC Name 1-méthyl-3-(trifluorométhyl)pyrazole-4-carboxylique acide
SMILES CN1C=C(C(=N1)C(F)(F)F)C(=O)O

Spécifications

Melting Point 203°C
Color Blanc
Quantity 1 g
Formula Weight 194.11
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)pyrazole-4-carboxylic Acid
EINECSNumber (5)-6995
TSCA No