missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™

Catalog No. AW00087DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AW00087DA 1g Amber Glass Bottle
AW00087DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AW00087DA Supplier Thermo Scientific Chemicals Supplier No. AW00087DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 519056-67-8
Molecular Formula C5H4ClF3N2O2S
Molecular Weight (g/mol) 248.604
InChI Key HJFWSNIHZZKYIK-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794561
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride