missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 879896-48-7
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.24
MDL Number MFCD08435884
InChI Key RKARRIJZFBGKSH-UHFFFAOYSA-N
Synonym 1-methyl-3-thien-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxaldehyde, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-2-thienyl, 2-methyl-5-thiophen-2-yl pyrazole-3-carbaldehyde, 1-methyl-3-2-thienyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl-pyrazole-5-carbaldehyde, 2-5-formyl-1-methyl-1h-pyrazol-3-yl thiophene
PubChem CID 18525809
IUPAC Name 2-methyl-5-thiophen-2-ylpyrazole-3-carbaldehyde
SMILES CN1N=C(C=C1C=O)C1=CC=CS1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC47204CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

879896-48-7
192.24
RKARRIJZFBGKSH-UHFFFAOYSA-N
18525809
CN1N=C(C=C1C=O)C1=CC=CS1
C9H8N2OS
MFCD08435884
1-methyl-3-thien-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxaldehyde, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-2-thienyl, 2-methyl-5-thiophen-2-yl pyrazole-3-carbaldehyde, 1-methyl-3-2-thienyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl-pyrazole-5-carbaldehyde, 2-5-formyl-1-methyl-1h-pyrazol-3-yl thiophene
2-methyl-5-thiophen-2-ylpyrazole-3-carbaldehyde

Specifications

879896-48-7
100.0
C9H8N2OS
1-methyl-3-thien-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxaldehyde, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-2-thienyl, 2-methyl-5-thiophen-2-yl pyrazole-3-carbaldehyde, 1-methyl-3-2-thienyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl-pyrazole-5-carbaldehyde, 2-5-formyl-1-methyl-1h-pyrazol-3-yl thiophene
CN1N=C(C=C1C=O)C1=CC=CS1
192.24
192.24
1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde
96.5
97%
MFCD08435884
RKARRIJZFBGKSH-UHFFFAOYSA-N
2-methyl-5-thiophen-2-ylpyrazole-3-carbaldehyde
18525809
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents