missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-propylimidazolium Bromide 98.0+%, TCI America™

Catalog No. M30345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30345G 5 g
M303425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30345G Supplier TCI America Supplier No. M30345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85100-76-1
Molecular Formula C7H13BrN2
MDL Number MFCD17014727

Specifications

Quantity 5 g
Formula Weight 205.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-propylimidazolium Bromide