missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$124.89 - $367.22
Chemical Identifiers
| CAS | 216299-72-8 |
|---|---|
| Molecular Formula | C9H13F6N3O4S2 |
| Molecular Weight (g/mol) | 405.33 |
| MDL Number | MFCD15071469 |
| InChI Key | CDWUIWLQQDTHRA-UHFFFAOYSA-N |
| PubChem CID | 12040508 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-methyl-3-propylimidazol-1-ium |
| SMILES | CCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 216299-72-8 | |
| 405.33 | |
| CDWUIWLQQDTHRA-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-3-propylimidazol-1-ium |
| C9H13F6N3O4S2 | |
| MFCD15071469 | |
| 12040508 | |
| CCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Specifications
| 216299-72-8 | |
| C9H13F6N3O4S2 | |
| 5 g | |
| CCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 405.33 | |
| 405.33 | |
| Liquid |
| Yellow | |
| MFCD15071469 | |
| CDWUIWLQQDTHRA-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-3-propylimidazol-1-ium | |
| 12040508 | |
| ≥98.0% (HPLC,T) | |
| 1-Methyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide |
Safety and Handling
TSCA : No