missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenylpiperazine 98.0+%, TCI America™

Catalog No. M16855G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16855G Supplier TCI America Supplier No. M16855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5271-27-2
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
InChI Key IRMBVBDXXYXPEW-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-piperazine, 1-n-methyl-3-phenyl-piperazine, 1-methyl-3-phenyl piperazine, piperazine, 1-methyl-3-phenyl, +/--3-phenyl-1-methylpiperazine, dl-1-methyl-3-phenyl-piperazine, 1-methyl-3-phenyl-1-piperazine, pubchem8578, 3-phenyl-1-methylpiperazine, +/-, n-methyl-3-phenylpiperazine
PubChem CID 2760009
IUPAC Name 1-methyl-3-phenylpiperazine
SMILES CN1CCNC(C1)C2=CC=CC=C2

Specifications

Melting Point 60°C
Color Yellow
pH 11.0 to 12.0 (5% H2O 25°C)
Quantity 5 g
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3-phenylpiperazine