missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenylpiperazine, 97%

Catalog No. AAH2692403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2692403 1 g
AAH2692406 5 g
AAH2692414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2692403 Supplier Thermo Scientific Chemicals Supplier No. H2692403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5271-27-2 | C11H16N2 | 176.263 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5271-27-2
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD03411603
InChI Key IRMBVBDXXYXPEW-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-piperazine, 1-n-methyl-3-phenyl-piperazine, 1-methyl-3-phenyl piperazine, piperazine, 1-methyl-3-phenyl, +/--3-phenyl-1-methylpiperazine, dl-1-methyl-3-phenyl-piperazine, 1-methyl-3-phenyl-1-piperazine, pubchem8578, 3-phenyl-1-methylpiperazine, +/-, n-methyl-3-phenylpiperazine
PubChem CID 2760009
IUPAC Name 1-methyl-3-phenylpiperazine
SMILES CN1CCNC(C1)C2=CC=CC=C2

Specifications

Melting Point 56°C to 60°C
Boiling Point 85°C (0.5 mmHg)
Quantity 1 g
UN Number UN2811
Sensitivity Air Sensitive
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-phenylpiperazine
Hazard Category H302+H312-H315-H318
Hazard Statement GHS H Statement
H318-H302-H312-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 432-360-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only