missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

$490.34 - $490.34

Chemical Identifiers

CAS 864068-96-2
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD08271941
InChI Key XFGHBJQGDDYIKS-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-phenylpyrazole-5-carbaldehyde, 2-methyl-5-phenyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-3-phenyl-1h-pyrazole, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-phenyl
PubChem CID 7537629
SMILES CN1N=C(C=C1C=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC46604DA
SDS View Documents
Thermo Scientific Chemicals
CC46604DA
1 g
Each for $490.34
Only null left
 
Description

Description

Specifications

Chemical Identifiers

864068-96-2
186.21
XFGHBJQGDDYIKS-UHFFFAOYSA-N
7537629
C11H10N2O
MFCD08271941
1-methyl-3-phenyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-phenylpyrazole-5-carbaldehyde, 2-methyl-5-phenyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-3-phenyl-1h-pyrazole, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-phenyl
CN1N=C(C=C1C=O)C1=CC=CC=C1

Specifications

864068-96-2
100.0
Amber glass bottle
MFCD08271941
1-methyl-3-phenyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-phenylpyrazole-5-carbaldehyde, 2-methyl-5-phenyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-3-phenyl-1h-pyrazole, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-phenyl
CN1N=C(C=C1C=O)C1=CC=CC=C1
186.21
186.21
1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde
96.5
97%
C11H10N2O
1 g
XFGHBJQGDDYIKS-UHFFFAOYSA-N
2-methyl-5-phenylpyrazole-3-carbaldehyde
7537629
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'