missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific™

Catalog No. p-3175576
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC46613DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC46613DA Supplier Thermo Scientific Chemicals Supplier No. CC46613DA

Chemical Identifiers

CAS 876728-39-1
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.25
MDL Number MFCD08271943
InChI Key XAVNJYHSYZUYFI-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazol-5-yl methylamine, 1-methyl-3-phenyl-1h-pyrazol-5-yl methanamine, 2-methyl-5-phenylpyrazol-3-yl methanamine, 1h-pyrazole-5-methanamine,1-methyl-3-phenyl, 1-1-methyl-3-phenyl-1h-pyrazol-5-yl methanamine, 1-methyl-3-phenylpyrazol-5-yl methylamine, 1-2-methyl-5-phenylpyrazol-3-yl methanamine, 5-aminomethyl-1-methyl-3-phenyl-1h-pyrazole, 5-aminomethyl-1-methyl-1h-pyrazol-3-yl benzene
PubChem CID 20109911
SMILES CN1N=C(C=C1CN)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (2-methyl-5-phenylpyrazol-3-yl)methanamine
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material (1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.