missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-nitroguanidine (wetted with ca. 25% Water) (unit weight on dry weight basis) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 160°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 1325 |
| Formula Weight | 118.10 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Methyl-3-nitroguanidine (wetted with ca. 25% Water) (unit weight on dry weight basis) |
Safety and Handling
| RTECSNumber | MF4125000 |
| TSCA | No |