missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-propyl-2-pyrazolin-5-one, 97%

Catalog No. p-7044243
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0542503 1 g
AAL0542506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0542503 Supplier Thermo Scientific Chemicals Supplier No. L0542503
Only null left

CAS: 31272-04-5 | C7H12N2O | 140.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31272-04-5
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00051849
InChI Key RULINAPACRUGOF-UHFFFAOYSA-N
Synonym 1-methyl-3-n-propyl-2-pyrazolin-5-one, 1-methyl-3-propyl-4,5-dihydro-1h-pyrazol-5-one, 1-methyl-3-propyl-1h-pyrazol-5 4h-one, 1-methyl-3-propyl-2-pyrazolin-5-one, 1-methyl-3-propyl-2-pyrazoline-5-one, 1-methyl-3-n-propyl 2-pyrazolin-5-one, 2-methyl-5-propyl-2,4-dihydro-3h-pyrazol-3-one, 3h-pyrazol-3-one,2,4-dihydro-2-methyl-5-propyl, 2-methyl-5-propyl-2,4-dihydro-3h-pyrazol-3-one #
PubChem CID 520516
IUPAC Name 2-methyl-5-propyl-4H-pyrazol-3-one
SMILES CCCC1=NN(C(=O)C1)C

Specifications

Melting Point 114°C to 118°C
Quantity 1 g
Beilstein 637013
Formula Weight 140.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-n-propyl-2-pyrazolin-5-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only