missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. M27325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27325G 5 g
M273225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27325G Supplier TCI America Supplier No. M27325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 244193-52-0
Molecular Formula C12H23BF4N2
Molecular Weight (g/mol) 282.13
MDL Number MFCD03095434
InChI Key GXZCAMSPWNHTAE-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium tetrafluoroborate, 1-methyl-3-n-octylimidazolium tetrafluoroborate, 3-methyl-1-octylimidazolium tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, omimbf4, dsstox_cid_27919, dsstox_rid_82672, dsstox_gsid_47943, 3-methyl-1-octylimidazoliumtetrafluoroborate, 1-methyl-3-octylimidazolium tetrafluoroborate hplc
PubChem CID 11460221
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Quantity 5 g
Formula Weight 282.13
Percent Purity ≥97.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Tetrafluoroborate