missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-indolemethylamine, 96%

Catalog No. p-7032801
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
5 g
Catalog No. Quantity
AAH34418MD 250 mg
AAH3441806 5 g
2 options

Catalog No. AAH34418MD

Supplier: Thermo Scientific Chemicals H34418MD

Only null left

CAS: 19293-60-8 | C10H12N2 | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19293-60-8
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD06657101
InChI Key NOFZMDGMQKRLIV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-3-yl methanamine, 1-methylindol-3-yl methanamine, 1-methyl-3-indolemethylamine, 1-methyl-1h-indol-3-yl-methylamine, 1-methyl-1h-indol-3-yl methylamine, 1-1-methyl-1h-indol-3-yl methanamine, c-1-methyl-1h-indol-3-yl-methylamine, 1-1-methylindol-3-yl methanamine, n-methyl-3-aminomethyl-indol, 1-methyl-1h-indole-3-methanamine
PubChem CID 3934542
IUPAC Name (1-methylindol-3-yl)methanamine
SMILES CN1C=C(C2=CC=CC=C21)CN

Specifications

Quantity 250 mg
Formula Weight 160.22
Percent Purity 96%
Chemical Name or Material 1-Methyl-3-indolemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only