missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-indolemethylamine, 96%

Catalog No. AAH34418MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34418MD 250 mg
AAH3441806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34418MD Supplier Thermo Scientific Chemicals Supplier No. H34418MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19293-60-8 | C10H12N2 | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19293-60-8
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD06657101
InChI Key NOFZMDGMQKRLIV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-3-yl methanamine, 1-methylindol-3-yl methanamine, 1-methyl-3-indolemethylamine, 1-methyl-1h-indol-3-yl-methylamine, 1-methyl-1h-indol-3-yl methylamine, 1-1-methyl-1h-indol-3-yl methanamine, c-1-methyl-1h-indol-3-yl-methylamine, 1-1-methylindol-3-yl methanamine, n-methyl-3-aminomethyl-indol, 1-methyl-1h-indole-3-methanamine
PubChem CID 3934542
IUPAC Name (1-methylindol-3-yl)methanamine
SMILES CN1C=C(C2=CC=CC=C21)CN

Specifications

Quantity 250 mg
Formula Weight 160.22
Percent Purity 96%
Chemical Name or Material 1-Methyl-3-indolemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only